ENA428074909 Enamine

methyl 3-isocyanato-3-methylbutanoate

  • MDL number MFCD24450447
  • Cas Number : 50835-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-182830
Availability details for
Quantity
  • Availability
  • Add to Cart