ENA428074928 Enamine

3-amino-N-(5-bromo-2-hydroxyphenyl)-4-methylbenzamide

  • MDL number MFCD24928001
  • Cas Number : 1525908-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182586
Availability details for
Quantity
  • Availability
  • Add to Cart