ENA428074941 Enamine

3-(iodomethyl)oxetane

  • MDL number MFCD18909284
  • Cas Number : 1003013-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-182629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182629
Availability details for
Quantity
  • Availability
  • Add to Cart