ENA428074944 Enamine

tert-butyl N-(2,2-dimethyl-3-oxopropyl)carbamate

  • MDL number MFCD12031785
  • Cas Number : 195387-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183860
Availability details for
Quantity
  • Availability
  • Add to Cart