ENA428075038 Enamine

tert-butyl N-(2-formylcyclohexyl)carbamate

  • MDL number MFCD24391095
  • Cas Number : 1496985-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-183866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183866
Availability details for
Quantity
  • Availability
  • Add to Cart