ENA428075040 Enamine

1-(phenylsulfonyl)octahydro-1H-indole-2-carboxylic acid

  • MDL number MFCD28063782
  • Cas Number : 1009251-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26615
Availability details for
Quantity
  • Availability
  • Add to Cart