ENA428075045 Enamine

4-[(4-methoxyphenyl)carbonyl]piperidine hydrochloride

  • MDL number MFCD00114817
  • Cas Number : 25519-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 257...259
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30360

Safety Information

signalword Warning
safety_statement 53-23-36/37/39-45
msds https://www.enaminestore.com/catalog/EN300-30360
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart