ENA428075049 Enamine

4-phenyl-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD24390314
  • Cas Number : 499216-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49286
Availability details for
Quantity
  • Availability
  • Add to Cart