ENA428075051 Enamine

2,4-diiodo-6-nitroaniline

  • MDL number MFCD03931845
  • Cas Number : 116529-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54335

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54335
Availability details for
Quantity
  • Availability
  • Add to Cart