ENA428075053 Enamine

octahydroindolizine

  • MDL number MFCD02066234
  • Cas Number : 13618-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61515
Availability details for
Quantity
  • Availability
  • Add to Cart