ENA428075054 Enamine

2,2-dimethyl-2,3-dihydro-1-benzofuran

  • MDL number MFCD12028054
  • Cas Number : 6337-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61932

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61932
Availability details for
Quantity
  • Availability
  • Add to Cart