ENA428075057 Enamine

2-phenyl-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD00451771
  • Cas Number : 1767-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292-293
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65425
Availability details for
Quantity
  • Availability
  • Add to Cart