ENA428075059 Enamine

1-benzoyl-octahydro-1H-indole-2-carboxylic acid

  • MDL number MFCD09044778
  • Cas Number : 120084-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66594
Availability details for
Quantity
  • Availability
  • Add to Cart