ENA428075068 Enamine

methyl 2-methylidenebutanoate

  • MDL number MFCD25542305
  • Cas Number : 2177-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86241
Availability details for
Quantity
  • Availability
  • Add to Cart