ENA428075073 Enamine

4-(hydroxymethyl)piperidin-3-ol

  • MDL number MFCD09036027
  • Cas Number : 220218-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95639
Availability details for
Quantity
  • Availability
  • Add to Cart