ENA428682410 Enamine

(2R,4S)-1-acetyl-4-methoxypyrrolidine-2-carboxylic acid

  • MDL number MFCD28118206
  • Cas Number : 1807885-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158272
Availability details for
Quantity
  • Availability
  • Add to Cart