ENA428682424 Enamine

3-tert-butyl-2,3-dihydro-1H-indole

  • MDL number MFCD28118241
  • Cas Number : 1692219-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176523
Availability details for
Quantity
  • Availability
  • Add to Cart