ENA428682430 Enamine

2-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-1,3-thiazole

  • MDL number MFCD28118251
  • Cas Number : 1803586-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177305
Availability details for
Quantity
  • Availability
  • Add to Cart