ENA428682431 Enamine

1-(4-bromobenzenesulfonyl)piperidin-3-amine hydrochloride

  • MDL number MFCD28118252
  • Cas Number : 1803592-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177489
Availability details for
Quantity
  • Availability
  • Add to Cart