ENA428682432 Enamine

2-[(2-aminoethyl)(methyl)amino]-N,N-dimethylacetamide dihydrochloride

  • MDL number MFCD28118253
  • Cas Number : 1803602-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177509
Availability details for
Quantity
  • Availability
  • Add to Cart