ENA428682433 Enamine

(2-aminoethyl)bis(2-methoxyethyl)amine dihydrochloride

  • MDL number MFCD28118254
  • Cas Number : 1803581-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177511
Availability details for
Quantity
  • Availability
  • Add to Cart