ENA428682442 Enamine

3,3-dimethyl-2-(prop-2-en-1-yl)piperidine

  • MDL number MFCD28118268
  • Cas Number : 1803595-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178644
Availability details for
Quantity
  • Availability
  • Add to Cart