ENA428682443 Enamine

3-(benzyloxy)-5-(chloromethyl)pyridine hydrochloride

  • MDL number MFCD28118271
  • Cas Number : 1803605-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178821
Availability details for
Quantity
  • Availability
  • Add to Cart