ENA428682445 Enamine

methyl 2-[5-(difluoromethyl)-1,3,4-oxadiazol-2-yl]acetate

  • MDL number MFCD28118274
  • Cas Number : 1798713-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179132
Availability details for
Quantity
  • Availability
  • Add to Cart