ENA428682449 Enamine

1-ethyl-N-methyl-2-(1-methyl-1H-pyrazol-4-yl)-6-oxopiperidine-3-carboxamide

  • MDL number MFCD28118280
  • Cas Number : 1803570-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179542
Availability details for
Quantity
  • Availability
  • Add to Cart