ENA428682450 Enamine

ethyl 3-(propan-2-yl)pyrrolidine-3-carboxylate

  • MDL number MFCD28118281
  • Cas Number : 1692722-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179802
Availability details for
Quantity
  • Availability
  • Add to Cart