ENA428682453 Enamine

3-amino-2-[(3,4,5-trifluorophenyl)methyl]propanoic acid hydrochloride

  • MDL number MFCD28118285
  • Cas Number : 1798747-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180003
Availability details for
Quantity
  • Availability
  • Add to Cart