ENA428682454 Enamine

4-(2,3-dihydro-1,4-benzodioxine-6-carbonyl)piperidine hydrate hydrochloride

  • MDL number MFCD28118286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl, H2O
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180027
Availability details for
Quantity
  • Availability
  • Add to Cart