ENA428682459 Enamine

3-(aminomethyl)-6-methyl-1,2-dihydropyridin-2-one dihydrochloride

  • MDL number MFCD28118291
  • Cas Number : 857429-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180282
Availability details for
Quantity
  • Availability
  • Add to Cart