ENA428682460 Enamine

3-amino-2-[(3-bromophenyl)methyl]propan-1-ol hydrochloride

  • MDL number MFCD28118292
  • Cas Number : 1803606-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180337
Availability details for
Quantity
  • Availability
  • Add to Cart