ENA428682471 Enamine

{4-[(3-methylphenyl)methoxy]phenyl}methanamine hydrochloride

  • MDL number MFCD28118308
  • Cas Number : 1803606-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180626
Availability details for
Quantity
  • Availability
  • Add to Cart