ENA428682472 Enamine

1-(2-aminoethyl)cyclopentan-1-ol hydrochloride

  • MDL number MFCD28118309
  • Cas Number : 859318-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180645
Availability details for
Quantity
  • Availability
  • Add to Cart