ENA428682476 Enamine

2-(2,4-dimethylpiperidin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28118318
  • Cas Number : 1803606-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180938
Availability details for
Quantity
  • Availability
  • Add to Cart