ENA428682479 Enamine

2-(3-ethylpiperidin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28118321
  • Cas Number : 1803599-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181012
Availability details for
Quantity
  • Availability
  • Add to Cart