ENA428682481 Enamine

ethyl 2-[(5-chloro-6-oxo-1,6-dihydropyridazin-4-yl)sulfanyl]acetate

  • MDL number MFCD28118323
  • Cas Number : 1696195-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181191
Availability details for
Quantity
  • Availability
  • Add to Cart