ENA428682485 Enamine

5-(2-{6-chloro-[1,2,4]triazolo[4,3-b]pyridazin-3-yl}ethyl)-3-(2-methoxyphenyl)-1,2,4-oxadiazole

  • MDL number MFCD28118327
  • Cas Number : 1803571-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181332
Availability details for
Quantity
  • Availability
  • Add to Cart