ENA428682488 Enamine

ethyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)-1,3-thiazole-2-carboxylate

  • MDL number MFCD28118330
  • Cas Number : 1803608-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181371
Availability details for
Quantity
  • Availability
  • Add to Cart