ENA428682489 Enamine

ethyl 4-[3-(1,3-dioxaindan-5-yl)-1,2,4-oxadiazol-5-yl]-1,3-thiazole-2-carboxylate

  • MDL number MFCD28118331
  • Cas Number : 1803603-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181373
Availability details for
Quantity
  • Availability
  • Add to Cart