ENA428682493 Enamine

6-[3-(4-methoxyphenyl)-1H-pyrazol-1-yl]pyridazine-3-thiol

  • MDL number MFCD28118335
  • Cas Number : 1795515-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181382
Availability details for
Quantity
  • Availability
  • Add to Cart