ENA428682495 Enamine

5-[1-(pyrazin-2-yl)-1H-imidazol-4-yl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD28118337
  • Cas Number : 1803590-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181390
Availability details for
Quantity
  • Availability
  • Add to Cart