ENA428682497 Enamine

5-(chloromethyl)-N,N-dimethyl-1,3,4-oxadiazol-2-amine

  • MDL number MFCD28118340
  • Cas Number : 1692481-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181661
Availability details for
Quantity
  • Availability
  • Add to Cart