ENA428682501 Enamine

3-(3-bromo-5-methylphenyl)-2-cyanopropanamide

  • MDL number MFCD28118356
  • Cas Number : 1803587-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182393
Availability details for
Quantity
  • Availability
  • Add to Cart