ENA428682505 Enamine

2-methyl-2,6-diazaspiro[3.4]octane dihydrochloride

  • MDL number MFCD28118361
  • Cas Number : 1795283-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182455
Availability details for
Quantity
  • Availability
  • Add to Cart