ENA428682508 Enamine

5-azaspiro[3.4]octane hydrobromide

  • MDL number MFCD28118364
  • Cas Number : 1795283-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182532
Availability details for
Quantity
  • Availability
  • Add to Cart