ENA428682509 Enamine

4-(trifluoromethyl)piperidin-4-amine dihydrochloride

  • MDL number MFCD28118365
  • Cas Number : 1803560-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182559
Availability details for
Quantity
  • Availability
  • Add to Cart