ENA428682512 Enamine

1-(5-fluoropyridin-3-yl)piperazine dihydrochloride

  • MDL number MFCD28118368
  • Cas Number : 1803560-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182628
Availability details for
Quantity
  • Availability
  • Add to Cart