ENA428682514 Enamine

1-(aminomethyl)-3-ethoxycyclobutan-1-ol

  • MDL number MFCD28118370
  • Cas Number : 1699420-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182648
Availability details for
Quantity
  • Availability
  • Add to Cart