ENA428682518 Enamine

(3S,5R)-5-(3-methyl-1,2,4-oxadiazol-5-yl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD28118374
  • Cas Number : 1820581-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182665
Availability details for
Quantity
  • Availability
  • Add to Cart