ENA428682521 Enamine

ethyl 2-(propan-2-yl)oxane-3-carboxylate

  • MDL number MFCD28118377
  • Cas Number : 1795499-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182702
Availability details for
Quantity
  • Availability
  • Add to Cart