ENA428682531 Enamine

3-methanesulfonylazepane hydrochloride

  • MDL number MFCD28118391
  • Cas Number : 1788990-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182923
Availability details for
Quantity
  • Availability
  • Add to Cart